FLUORESCENCE SEPARATION MODULE 2-40 KDA
Catalog # MSPP-SM-FL006
Supplier: PROTEIN SIMPLE MS
FLUORESCENCE SEPARATION MODULE 2-40 KDA
Catalog # MSPP-SM-FL006
Supplier: PROTEIN SIMPLE MS
Supplier Number: SM-FL006
Frequently Bought Together
ROCK SALT
Supplier: INDUSTRIAL SUPPORT INC.
PROTEIN NORMALIZATION MODULE
Supplier: 0000014126
DETECTION MODULE ANTI-MOUSE W/PS/SS
Supplier: PROTEIN SIMPLE MS
SEPARATION MODULE CARTRID 2X13 2-40 KDA
Supplier: 0000014126
Anti-Rabbit Detection Module, Protein Simple
Supplier: PROTEIN SIMPLE MS
TOTAL PROTEIN DETECTION MODULE
Supplier: 0000014126
EZ STANDARD PACK 1 PK8
Supplier: 0000014126
MODULE ANTI-RABBIT IR DETECTION
Supplier: PROTEIN SIMPLE MS
STANDARD EZ WITH 8 PACKETS 66-440KDA PK3
Supplier: PROTEIN SIMPLE MS
Jess or Wes Separation Module, Protein Simple
Supplier: PROTEIN SIMPLE MS
ANTIBODY GOAT ANTI-MOUSE HRP CONJUGATE
Supplier: PROTEIN SIMPLE MS
CONJUGATE 20X ANTI-RABBIT HRP 150UL
Supplier: PROTEIN SIMPLE MS
WASH BUFFER 60ML
Supplier: 0000014126
KIT EZ STANDARD PACK 4 PK8
Supplier: 0000014126
BioProfile FLEX 2 Calibrator Cartridge LV, Nova Biomedical
Supplier: Nova Biomedical
ANTIBODY ANTI-RABBIT SECONDARY
Supplier: PROTEIN SIMPLE MS
SEPARATION MODULE WES 66-440KDA 8X25 CC
Supplier: 0000014126
KIT REPLEX MODULE FOR JESS
Supplier: 0000014126
BioProfile® Flex 2 Microsensor Cards for Automated Chemistry Analyzers, Nova Biomedical
Supplier: Nova Biomedical
DILUENT MILK-FREE ANTIBODY 20ML
Supplier: 0000014126