SOLUTION TRYPSIN NEUTRALIZING 100ML
Catalog # MSPP-PCS9990
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
SOLUTION TRYPSIN NEUTRALIZING 100ML
Catalog # MSPP-PCS9990
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: PCS-999004BSL1
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Cell Culture Plates Treated for Increased Cell Attachment, Sterile
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Universal Pipette Tips
Supplier: VWR International
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International