Orders LinkContactUsLinkComponent
Crazy Circuits Classroom Sets: Programming 101
Crazy Circuits Classroom Sets: Programming 101
Crazy Circuits: Programming 101, 25 packs, 1 teacher box + 24 student boxes
Catalog # 470348-922
Supplier #:  K1051-25PK
Crazy Circuits Classroom Sets: Programming 101
Catalog # 470348-922
Supplier:  
Crazy Circuits Classroom Sets: Programming 101
Crazy Circuits Classroom Sets: Programming 101
Crazy Circuits: Programming 101, 25 packs, 1 teacher box + 24 student boxes
Discontinued
Product Details & Documents
A complete classroom solution to the daunting task of teaching electronics and programming.
  • As with all Crazy Circuits kits, our classroom sets require no soldering
  • Parts are connected in a brick-based building environment via our special maker tape conductive tape
  • The only additional tools you'll need are scissors and a computer to manipulate code
The Programming 101 Classroom line was designed as a complete classroom solution for teaching electronics. These modular kits are robust sets of materials supported by online lesson plans, projects, activities, diagrams, and videos. Programming 101 sets each come with one Teacher box and your choice of 4, 9, or 24 Student boxes filled with electronics parts and supplies. The Teacher box includes a full set of Student supplies as well as additional materials. These modular kits are supported by equally robust, NGSS-aligned, online teaching resources designed to facilitate easy classroom use. In addition to lesson plans, the online resources also include activity pages, assessments, diagrams, and videos for classroom use.
Ordering information: Scissors and computer(s) are required but not included.
Delivery information: Each Student box includes 42 Parts + organizer, (5) Switches (pushbutton, slide), (4) LEDs, (4) Resistors, (4) Brick plates, (3) Ribbon wire sets, (2) Female headers, (2) Potentiometers, (1) Roll ⅛” Maker Tape™, (1) Photoresistor, (1) Neopixel, (1) Neopixel strip, (1) Programmable robotics (Arduino™) board, (1) LEGO™ compatible servo + connector, (1) Ultrasonic distance sensor, (1) 4-Bit 7 segment display, (1) Joystick, (1) Temperature/humidity sensor, (1) Brick baseplate, (1) USB cable, (1) Bundle of 5 alligator clips, Instructions cards/online curriculum. Teacher box includes (1) Complete contents of Student box, (1) Large organizer (replaces organizer from Student box), (4) additional rolls of ⅛" Nylon conductive tape, (2) Additional female to female ribbon wire sets.
Specifications
  • Description:
    Crazy Circuits: Programming 101, 25 packs
  • Includes:
    1 teacher box + 24 student boxes