EP Reference Solution, Brown Reference Solution, RICCA Chemical Company
EP Reference Solution B5, Brown Reference Solution, 1 L
Supplier: Ricca Chemical
Supplier #: R2880500-1C
EP Reference Solution, Brown Reference Solution, RICCA Chemical Company
Catalog # RCR2880500-1C
Supplier: Ricca Chemical
CAS Number:
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:EP Reference Solution B5, Brown Reference Solution
- Size:1 L
- Cat. No.:RCR2880500-1C
- Appearance:Brown liquid
- Storage Temperature:Ambient
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



