Orders LinkContactUsLinkComponent
Life/form® Basic Sanitary CPR Dog
Life/form® Basic Sanitary CPR Dog
Dog CPR, advanced
Catalog # 470348-734
Supplier:   Nasco
Supplier #:  LF01155
Life/form® Basic Sanitary CPR Dog
Catalog # 470348-734
Supplier:  Nasco
Life/form® Basic Sanitary CPR Dog
Life/form® Basic Sanitary CPR Dog
Dog CPR, advanced
Price
$2,668.85
Log in for availability
Each
Product Details & Documents
Learn canine CPR.
  • Basic and advanced options
  • Muzzles can be disinfected
  • 5-year warranty
The Life/form® basic sanitary CPR dog provides each student with their own sanitary muzzle and nosepiece with a one-way valve. The muzzles may be disinfected and reused, while the nosepieces and lower airways are disposable.
In addition to practice in assisted breathing, students may practice the appropriate rate, pressure, and position of cardiac massage and coordinate the respiratory cardiovascular functions. A visible chest rise appears when correct ventilations are applied. Includes soft carrying case, six muzzles, 10 nosepieces, and three lower disposable airways.
The Life/form® advanced sanitary CPR dog provides each student with their own sanitary muzzle and nosepiece with a one-way valve. The muzzles may be disinfected and reused, while the nosepieces and lower airways are disposable. In addition to practice in assisted breathing, students may practice the appropriate rate, pressure, and position of cardiac massage and coordinate the respiratory cardiovascular functions.
An IV can even be established for more advanced training. The advanced CPR dog is connected to a small box that displays a light when the correct ventilation volume is applied, another light indicates correct position for cardiac massage, a third light indicates correct compression depth, and an audible beep and light indicates excessive pressure is being applied. A visible chest rise appears when correct ventilations are applied.
Delivery information: Includes 10 disposable nosepieces, three disposable lower airways, endotracheal tube, lubricant, 12 cc syringe, towels, butterfly set, artificial blood, two IV bags, control box with AC adapter, 9V battery, and soft carry case.
Specifications
  • Description:
    Dog CPR, advanced