Orders LinkContactUsLinkComponent
Simulaids® Catheterization Trainer
Simulaids® Catheterization Trainer
Male trainer
Catalog # 470348-662
Supplier:   Nasco
Supplier #:  160-1931
Simulaids® Catheterization Trainer
Catalog # 470348-662
Supplier:  Nasco
Simulaids® Catheterization Trainer
Simulaids® Catheterization Trainer
Male trainer
Price
$523.15
Log in for availability
Each
Product Details & Documents
Newly designed anatomically correct models.
  • Available in male, female, or a set of both
  • Anatomically correct
  • Reservoir provides urine flow when successful
Training is enhanced by a new material that has a realistic feel and flexibility. Both models utilize a 16 FR catheter and can be used to demonstrate balloon inflation procedures. The models are individually mounted on a training stand and can be used on a tabletop or for patient simulation.
A fluid reservoir allows for urine output upon successful completion of the insertion procedure. Instructors are able to teach sterile technique of insertion and removal of catheter, as well as practice preparing the patient for the procedure.
Specifications
  • Description:
    Male trainer