Orders LinkContactUsLinkComponent
Accessories for CR2700 Barcode Scanner
Accessories for CR2700 Barcode Scanner
Dongle, 1.7H×0.73W×0.04"D, CR2700
Catalog # 76637-566
Supplier:   Brady Worldwide
Supplier #:  176513
Accessories for CR2700 Barcode Scanner
Catalog # 76637-566
Supplier:  Brady Worldwide
Accessories for CR2700 Barcode Scanner
Accessories for CR2700 Barcode Scanner
Dongle, 1.7H×0.73W×0.04"D, CR2700
Price
$207.61
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
CR2700 Barcode Scanner USB cables, chargers, power supplies, and mounting brackets.
Keep the CR2700 charged with this inductive charging station. Inductive charging eliminates exposed metal and related contact failure caused by chemical wipe-downs. Bluetooth capability wirelessly connects the CR2700 to a computer or a printer via the charging station. Code US Power Supply, 100 Watts.
A 3' straight USB cable to make a USB to Micro USB connection. Reader termination: Micro USB. Host termination: USB Type A. A 3' straight USB cable to make a USB to Micro USB connection. Reader termination: Micro USB. Host termination: USB Type A.
Specifications
  • Description:
    Dongle, 1.7H×0.73W×0.04"D
  • For model:
    CR2700