Orders LinkContactUsLinkComponent
Motic AE2000 Inverted Microscope LED
Motic AE2000 Inverted Microscope LED
AE2000 trinocular inverted microscope LED, basic package
Catalog # 76595-446
Supplier:   Motic
Supplier #:  1100103800266
Motic AE2000 Inverted Microscope LED
Catalog # 76595-446
Supplier:  Motic
Motic AE2000 Inverted Microscope LED
Motic AE2000 Inverted Microscope LED
AE2000 trinocular inverted microscope LED, basic package
Price
$3,444.70
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
The AE2000 is a revolutionary design that is packed with features and sets a new benchmark in affordable inverted microscopy. Motic’s AE2000 inverted microscope is designed for routine live cell inspection with an easy and flexible set up, ideal for university, routine-clinical, and lab environments.
  • Head Siedentopf type 45° inclined
  • Side facing quadruple nosepiece
  • CCIS plan achromat objective PL PH4X, PL PH 10X (basic package)
  • Optional CCIS plan achromat objective with PL PH 20X
  • ELWD N.A. 0.30 condenser
  • Phase slider Ph0, BDF, Ph1
  • Centering telescope
  • Plain stage with metal and glass stage inserts
  • 3 W LED illumination with intensity control
Binocular inverted live cell microscope for routine microbiology. The pre-defined optical setup with LWD CCIS© Phase objectives is ready for the inspection of diverse native samples in cell culture flasks. Lab technicians benefit from the convincing phase contrast concept, while the Fixed-Köhler illumination with interchangeable light source (HAL/LED) and auto ON/OFF function enables a fast throughput of samples.
Ordering information: Contact your Avantor representative to learn more about this and other motic solutions. All motic products come with a one-year electrical and five-year mechanical warranty.
Specifications
  • Description:
    AE2000 trinocular inverted microscope LED, basic package
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Glass stage insert
    Glass stage insert
    89168-420
    Log in for availability
    Each - $28.50
Product Information
  • Catalog #
    Availability
    Price
  • CCIS Plan Achromat Phase PL Ph4x, N.A. 0.10 / W.D. 12.6, Ph0
    CCIS Plan Achromat Phase PL Ph4x, N.A. 0.10 / W.D. 12.6, Ph0
    89168-402
    Log in for availability
    Each - $188.51
Product Information
  • Catalog #
    Availability
    Price
  • CCIS Plan Achromat LWD PL 20X N.A.0.3 / W.D. 4.7
    CCIS Plan Achromat LWD PL 20X N.A.0.3 / W.D. 4.7
    89168-398
    Log in for availability
    Each - $288.24
Product Information
  • Catalog #
    Availability
    Price
  • Widefield Eyepiece N-WF 15x/16mm
    Widefield Eyepiece N-WF 15x/16mm
    89168-442
    Log in for availability
    Each - $150.14
Product Information
  • Catalog #
    Availability
    Price
  • Phase Stop Ph0 (PL Ph4x)
    Phase Stop Ph0 (PL Ph4x)
    89168-414
    Log in for availability
    Each - $52.61
Product Information
  • Catalog #
    Availability
    Price
  • Hemacytometer holder
    Hemacytometer holder
    19000-688
    Log in for availability
    Each - $78.38
Product Information
  • Catalog #
    Availability
    Price
  • 0.5x C-Mount Camera Adapter (for 0.5" Chip Sensors)
    0.5x C-Mount Camera Adapter (for 0.5" Chip Sensors)
    10767-528
    Log in for availability
    Each - $185.54
Product Information
  • Catalog #
    Availability
    Price
  • CCIS Plan Achromat LWD PL 40X N.A. 0.5 / W.D. 3
    CCIS Plan Achromat LWD PL 40X N.A. 0.5 / W.D. 3
    89168-400
    Log in for availability
    Each - $389.04
Product Information
  • Catalog #
    Availability
    Price
  • Phase slider, centerable, BF, centerable
    Phase slider, centerable, BF, centerable
    89168-412
    Log in for availability
    Each - $192.88
Product Information
  • Catalog #
    Availability
    Price
  • 0.65x C-Mount Camera Adapter (for 0.66" Chip Sensors)
    0.65x C-Mount Camera Adapter (for 0.66" Chip Sensors)
    10767-530
    Log in for availability
    Each - $202.45
Product Information
  • Catalog #
    Availability
    Price
  • 1x C-Mount Camera Adapter (no Lens)
    1x C-Mount Camera Adapter (no Lens)
    10767-532
    Log in for availability
    Each - $192.82
Product Information
  • Catalog #
    Availability
    Price
  • Phase Stop Ph1 (PL Ph 10x, PL Ph 20x)
    Phase Stop Ph1 (PL Ph 10x, PL Ph 20x)
    89168-416
    Log in for availability
    Each - $52.61
Product Information
  • Catalog #
    Availability
    Price
  • CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8
    CCIS Plan Achromat PL 10X N.A. 0.25 / W.D. 16.8
    89168-396
    Log in for availability
    Each - $188.51