CARTRIDGE CHISEL JBC C115108 0.6 X 0.3
Catalog # MSPP-C115108
Supplier: TEQUIPMENT.NET MS
CARTRIDGE CHISEL JBC C115108 0.6 X 0.3
Catalog # MSPP-C115108
Supplier: TEQUIPMENT.NET MS
Supplier Number: JBCC115108
Frequently Bought Together
Dispensing Tips, EFD INC.
Supplier: EFD INC.- CUSTOMER SERVICE TE
UltraSOLV® Cleaning Swabs, Foamtec
Supplier: FOAMTEC INTERNATIONAL, LLC.
NANO HANDLE 103X14X14MM 1.2M
Supplier: JBC TOOLS USA INC. TE
Orion™ Star™ A214 pH/ISE Benchtop Meter, Thermo Scientific
Supplier: Thermo Fisher Scientific
CARTRIDGE CONICAL DIA 0.3 S1 SOLDERING
Supplier: JBC TOOLS USA INC. TE
Orion™ Versa Star Pro™ pH/ISE Benchtop Multiparameter Meter
Supplier: Thermo Fisher Scientific
12-14 HEALTHCARE CAT CASE
Supplier: VWR International
BPTES >97% POWDER 314045-39-1 10MG
Supplier: Selleck Chemicals
Disodium oxalate 99%
Supplier: Ambeed
MAGNA AMP LOG 6 PK50
Supplier: MESA LABS MS
Set of Casters for 6 Gallon Buckets, Vileda Professional®
Supplier: Vileda Professional - FHP
EDTA tripotassium salt dihydrate 99+%
Supplier: Thermo Scientific Chemicals
Soldering Tips for NP105 and NT105 Nano Tools, JBC
Supplier: JBC TOOLS USA INC. TE
Octyltriethoxysilane 97%
Supplier: Ambeed
Assurance® Grade Manganese Standards for AA and ICP, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
Fluid Thioglycollate Medium, Dehydrated, HiMedia Laboratories
Supplier: HiMedia
Benzylamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Sabdex (Sabouraud Dextrose) Agar
Supplier: Hardy Diagnostics