KIT ICELL CARDIOMYOCYTES CRYOPR 5X10 6V
Catalog # MSPP-R1017
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
KIT ICELL CARDIOMYOCYTES CRYOPR 5X10 6V
Catalog # MSPP-R1017
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
Supplier Number: R1017
Frequently Bought Together
Alcohol reagent ≥70% ACS
Supplier: Avantik
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
TIP FILTER LOW STERILE 125 UL XYZ CS1920
Supplier: 0000018277
PLATE CYTOVIEW MEA BLACK 48WELL
Supplier: AXION BIOSYSTEMS, INC MS
U-SLIDE 8 WELL HIGH GLASS BOTTOM PK15
Supplier: IBIDI USA MS
Corning® Matrigel® Basement Membrane Matrix, Corning
Supplier: Corning
EarlyTox™ Cardiotoxicity Kit
Supplier: MOLECULAR DEVICES, LLC MS
Jess or Wes Separation Module, Protein Simple
Supplier: PROTEIN SIMPLE MS
RACKS OF 384 TIPS 12.5 L 5 XYZ STERILE
Supplier: 0000018277
RNACLEAN XP LIQUID 40 ML
Supplier: Beckman Coulter
MEDIUM CULTURE CARDIOMYOCYTE 250ML
Supplier: NCARDIA BV MS
ICELL HEPATOCYTES 2.0 01279
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
ACONITINE 97%, 25MG
Supplier: Pfaltz & Bauer
CARDIOMYOCYTES ICELL 4 X 106CELL/VIAL
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
Endothelial Cell Growth Medium MV, PromoCell
Supplier: Promocell, Inc.