Orders LinkContactUsLinkComponent
VWR® Cryo Coder Cap Inserts
VWR® Cryo Coder Cap Inserts
Cap insert, Tan
Catalog # VWRI210-000038
Supplier:   VWR International
VWR® Cryo Coder Cap Inserts
Catalog # VWRI210-000038
Supplier:  VWR International
VWR® Cryo Coder Cap Inserts
VWR® Cryo Coder Cap Inserts
Cap insert, Tan
Price
$35.93
New Customer Promotional Price
Log in for availability
Case of 500
Product Details & Documents
Product can be used as coding/identification of samples types stored in it.
Cryocoder/cap inserts are available in multiple color options: Violet, white, blue, orange, lilac, red, tan, yellow, green, grey, pink, assorted (five colours).
Specifications
  • Color:
    Tan
  • Description:
    Cap insert