KIT PROTEIN STAIN 520 WESTERN BOLT 500ML
Catalog # MSPP-926-10016
Supplier: Li-Cor
KIT PROTEIN STAIN 520 WESTERN BOLT 500ML
Catalog # MSPP-926-10016
Supplier: Li-Cor
Supplier Number: 926-10016
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
REVERT™ Total Protein Stain Kits, LI-COR®
Supplier: Li-Cor
Microcentrifuge Tube Tough-Tags, Thomas-Wiley
Supplier: Thomas Scientific
Anti-IgG Goat Polyclonal Antibody (IRDye® 680RD)
Supplier: Li-Cor
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International