CHROMAN 1 SOLID 5MG
Catalog # MSPP-HY-15392
Supplier: MEDCHEMEXPRESS LLC MS
CHROMAN 1 SOLID 5MG
Catalog # MSPP-HY-15392
Supplier: MEDCHEMEXPRESS LLC MS
Supplier Number: HY-15392
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
Puritan® Standard Cotton Tipped Applicators, Wood Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
BD Microtainer® Tubes, BD Diagnostics
Supplier: BD
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
NeoPro® Powder-Free Neoprene Gloves, Microflex®, Ansell
Supplier: Ansell Healthcare
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Bovine Serum Albumin (BSA)
Supplier: Jackson Immunoresearch Lab
EMRICASAN 99% CAS NUMBER 254750-02-2 5MG
Supplier: Selleck Chemicals
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
Supplier: Hardy Diagnostics
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Alkaline Phosphatase (APP) Chromogen Substrates for IHC, Vector Laboratories
Supplier: Vector Laboratories