Orders LinkContactUsLinkComponent
Sample Cuvettes
Sample Cuvettes
50 mm, Glass
Catalog # MSPP-2629250
Marketsource
Supplier:   HACH COMPANY MS
Supplier #:  2629250
Sample Cuvettes
Catalog # MSPP-2629250
Supplier:  HACH COMPANY MS
Marketsource
Sample Cuvettes
Sample Cuvettes
50 mm, Glass
Price
$412.62
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Sample cuvettes for photometric measurements.
  • Available with or without lid
  • Available reusable (made from glass) or for single use (made from plastics)
Specifications
  • Window material:
    Glass
  • Pathlength:
    50 mm