SALT INSTANT OCEAN AQUARIUM 50 GAL
MSPP-198140
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
VWR® Qualitative Filter Paper, Grade 413
: VWR International
Corning® Gosselin™ Petri Dishes, Corning
: Corning
Ward's® Marble (Fine White)
: Avantor
Acetone ≥99.5% ACS, VWR Chemicals BDH®
: VWR International
Brine Shrimp Eggs
: Avantor
VWR® Sterile 70% Isopropanol
: VWR International
Parafilm® M Sealing Film
: Bemis Company, Inc.
80-Well Micro Tube Racks, Bio Plas
: Bio Plas
VWR® Laboratory Wide Mouth Bottles, PP
: VWR International
Sol-M® Hypodermic Needles
: SOL-MILLENNIUM MEDICAL
Pacon® Easel Rolls
: Janitorial Supplies
Dysprosium, plasma standard solution, Specpure®, Dy 10000 µg/ml, Thermo Scientific
: Thermo Scientific Chemicals
Orthoboric acid ≥99.5%, powder NF
: Spectrum Chemicals