HPLC COLUMN SRT SEC 5UM 750A 7.8X300MM
Catalog # MSPP-215750-78
Supplier: Sepax Technologies
HPLC COLUMN SRT SEC 5UM 750A 7.8X300MM
Catalog # MSPP-215750-78
Supplier: Sepax Technologies
Supplier Number: 215750-7830
Frequently Bought Together
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Fused Silica Tubing Cutters for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® 11 mm Snap Ring Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
CrossLab Graphite/Vespel Ferrules for GC
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Inserts for High Recovery Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Gas Clean Purification System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab CE and CE/MS Filters and Frits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
GC Autosampler Syringes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
High Recovery Vials and Inserts, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Inland 45 Pump Oil, Restek
Supplier: Restek
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
Closures for 2 ml Screw Top Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
QuickTiter™ HIV Lentivirus Quantitation Kit (p24 ELISA), Cell Biolabs
Supplier: Cell Biolabs
HPLC COLUMN SRTSEC-750 5UM 750A 7.8X50MM
Supplier: Sepax Technologies
COLUMN SRT SRT SEC 5UM 750A 4.6X300MM
Supplier: Sepax Technologies