COLUMN MIDI TRANSFEC GR PLASMID DNA PK10
Catalog # MSPP-74041010S
Supplier: 0000014357
COLUMN MIDI TRANSFEC GR PLASMID DNA PK10
Catalog # MSPP-74041010S
Supplier: 0000014357
Supplier Number: 740410.10S
Frequently Bought Together
VWR® 11 mm Snap Ring Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR International
Closures for 2 ml Screw Top Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Disposable Pasteur Pipettes, Glass
Supplier: VWR International
CrossLab CE and CE/MS Filters and Frits, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® Pouring Ring for Bottles
Supplier: VWR International
VWR® Vacuum Filtration Systems, Standard Line
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Supplier: VWR International
OptiTDS™, Heart Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
MIDORI Green Advance Nucleic Acid Staining Solution, Bulldog-bio
Supplier: Bulldog Bio
L-Lactate Assay Kit, Cayman Chemical
Supplier: Cayman Chemical Company
BUFFER RESUPENSION RES 1000ML 15-25DEG C
Supplier: MACHEREY-NAGEL INC MS