BUFFER NEUTRALIZATION NEU EF 1000 ML
Catalog # MSPP-740388-1
Supplier: 0000014357
BUFFER NEUTRALIZATION NEU EF 1000 ML
Catalog # MSPP-740388-1
Supplier: 0000014357
Supplier Number: 740388.1000
Frequently Bought Together
Antibiotic Test Disc Sets
Supplier: Avantor
EZBio® Single-Use Assembly, Media Bottle, PETG, Vented with Tubing, Sterile, Foxx Life Sciences
Supplier: Foxx Life Sciences
Accessories for Aspirating Pipettes, Costar®, Corning
Supplier: Corning
PREempt™ RTU Disinfectant Wipes, Contec®
Supplier: Contec
Commode Specimen Collection System, Covidien
Supplier: Covidien
DxH 500 Hematology Analyzer, Beckman
Supplier: Beckman Coulter
BOND Dewax Solution, Leica Microsystem
Supplier: Leica Biosystems MS
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
BUFFER LYSIS LYS EF ENDOTOXIN 1000 ML
Supplier: MACHEREY-NAGEL INC MS
pH Buffers, YSI
Supplier: YSI
Osmolality Standards, Wescor®
Supplier: ELITECHGROUP INC. MS
Brushes, Red Sable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
BUFFER RESUSPENSION RES EF 1000 ML
Supplier: MACHEREY-NAGEL INC MS
ToughCut® Fine Scissors, Fine Science Tools
Supplier: 0000013460
DNA ScreenTape Analysis
Supplier: Agilent Technologies
FILTER CAPPED SPIN-AWAY W/MINIPREP PK50
Supplier: Zymo Research
Sample Vials for Vi-CELL BLU Analyzers
Supplier: Beckman Coulter