RECOMBINANT CYTOKINE CHO 90% 10UG
Catalog # MSPP-RCYCHILL2
Supplier: Invivogen
RECOMBINANT CYTOKINE CHO 90% 10UG
Catalog # MSPP-RCYCHILL2
Supplier: Invivogen
Supplier Number: RCYC-HIL12
Frequently Bought Together
L(+)-Cysteine hydrochloride monohydrate, high purity
Supplier: VWR
ROCK SALT
Supplier: INDUSTRIAL SUPPORT INC.
Benchtop DNA Marker
Supplier: PROMEGA SUNNYVALE INC SE
Ward's® Introduction to Agarose Gel Electrophoresis Lab Activity
Supplier: Avantor
12-14 HEALTHCARE CAT CASE
Supplier: VWR International
Sabdex (Sabouraud Dextrose) Agar
Supplier: Hardy Diagnostics
Using CRISPR to treat Cystic Fibrosis
Supplier: Edvotek
MAGNA AMP LOG 6 PK50
Supplier: MESA LABS MS
Fluid D Rinsing Solution, Biomerieux
Supplier: bioMerieux
INDICATOR CHEM STEAM PK=BX250
Supplier: Steris
Set of Casters for 6 Gallon Buckets, Vileda Professional®
Supplier: Vileda Professional - FHP
EDVOTEK HexagelTM M36 Electrophoresis Chamber
Supplier: Edvotek
Tryptic Soy Broth, Biomerieux
Supplier: bioMerieux
miniPCR® SeeGreen™ All-in-One Agarose Tabs™
Supplier: AMPLYUS LLC
PROTEIN SIZE MARKER WIDE VEIW PRESTAINED
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
ASSAY RPLEX HUMAN IL18BP K151J5R-2
Supplier: Meso Scale Diagnostics
Ward's® Improved Bacterial Transformation Using GFP Lab Activity
Supplier: Avantor