NEGATIVE CONTROL MOUSE READY TO USE
Catalog # MSPP-PA0996
Supplier: Leica Biosystems MS
NEGATIVE CONTROL MOUSE READY TO USE
Catalog # MSPP-PA0996
Supplier: Leica Biosystems MS
Supplier Number: PA0996
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International