Order Entry
United States
Orders LinkContactUsLinkComponent
Cole-Parmer® Essentials Mountable Cable Ties

Cole-Parmer® Essentials Mountable Cable Ties

18 pound nylon mountable cable/zip ties, White, 11.43 cm (4.5"), 2.2 cm (0.87") max. bundle Ø, 18 lbs.
Catalog # 76579-892
Supplier:   Antyila Scientific
Supplier #:  06842-52
Cole-Parmer® Essentials Mountable Cable Ties
Catalog # 76579-892
Supplier:  Antyila Scientific
Cole-Parmer® Essentials Mountable Cable Ties
Cole-Parmer® Essentials Mountable Cable Ties

18 pound nylon mountable cable/zip ties, White, 11.43 cm (4.5"), 2.2 cm (0.87") max. bundle Ø, 18 lbs.

Price
$7.01
New Customer Promotional Price
Log in for availability
Case of 100
Product Details & Documents

Mountable cable ties offer two products in one. Easily bundle cables together and permanently screw mount the ties to a wall, panel, or other surfaces using the integrated mounting eyelet on the cable tie head.

  • Requires no additional clips or mounts
  • Self-lock feature securely bundles wires and cables keeping them neat and organized

Ties are UL-component recognized, and are self-extinguishing as per UL 94 Flam. Class: V-2. Ties withstand temperatures to 85 °C (185 °F ) and have a melting point of 257 °C (495 °F).

Specifications
  • Ø:
    2.2 cm (0.87") max. bundle Ø
  • Color:
    White
  • Description:
    18 pound nylon mountable cable/zip ties
  • Length:
    11.43 cm (4.5")
  • Tensile strength:
    18 lbs.