BIO-GLO-NL LUCIFERASE ASSAY SYSTEM
Catalog # 76580-416
Supplier: Promega Corporation
BIO-GLO-NL LUCIFERASE ASSAY SYSTEM
Catalog # 76580-416
Supplier: Promega Corporation
Supplier Number: J3081
Frequently Bought Together
VWR® Mini Vortex Mixer
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
Specialty Cell Culture Media
Supplier: Lonza
[Leu27]-Melan-A/MART-1 (26-35)
Supplier: Anaspec Inc
MSD READ BUFFER T (4X) 200 ML
Supplier: Meso Scale Diagnostics