Orders LinkContactUsLinkComponent
Cole-Parmer® Essentials Mountable Cable Ties
40 pound nylon mountable cable/zip ties, Black, 15.24 cm (6"), 3.2 cm (1.26") max. bundle Ø, 40 lbs.
  76579-830
 :  06842-53
Cole-Parmer® Essentials Mountable Cable Ties
  76579-830
Cole-Parmer® Essentials Mountable Cable Ties
Cole-Parmer® Essentials Mountable Cable Ties
40 pound nylon mountable cable/zip ties, Black, 15.24 cm (6"), 3.2 cm (1.26") max. bundle Ø, 40 lbs.
$10.34
Case of 100
Mountable cable ties offer two products in one. Easily bundle cables together and permanently screw mount the ties to a wall, panel, or other surfaces using the integrated mounting eyelet on the cable tie head.
  • Requires no additional clips or mounts
  • Self-lock feature securely bundles wires and cables keeping them neat and organized
Ties are UL-component recognized, and are self-extinguishing as per UL 94 Flam. Class: V-2. Ties withstand temperatures to 85 °C (185 °F ) and have a melting point of 257 °C (495 °F).
  • Ø:
    3.2 cm (1.26") max. bundle Ø
  • Color:
    Black
  • Description:
    40 pound nylon mountable cable/zip ties
  • Length:
    15.24 cm (6")
  • Tensile strength:
    40 lbs.