Orders LinkContactUsLinkComponent
Heated Forceps, Mopec
Heated Forceps, Mopec
Heated forceps, Blue, Smooth, 4 mm
Catalog # 76579-236
Supplier:   Mopec
Supplier #:  BK304-SM
Heated Forceps, Mopec
Catalog # 76579-236
Supplier:  Mopec
Heated Forceps, Mopec
Heated Forceps, Mopec
Heated forceps, Blue, Smooth, 4 mm
Price
$697.79
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mopec’s heated forceps are electrically heated instruments designed to assist with the proper positioning of tissue specimens during the embedding process in paraffin wax. The safe, low-voltage heating element maintains a steady temperature throughout specimen manipulation.
  • Designed to take up minimal bench-top space
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Molded finger grips increase user comfort and remain cool when in use
  • Bayonet connection prevents disconnection while in use
The ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth. Choose between serrated or smooth tips. Available in three sizes and compatible with the Mopec control unit.
Specifications
  • Color:
    Blue
  • Description:
    Heated forceps
  • Tip style:
    Smooth
  • Tip width:
    4 mm