CELL LINE HT-1197 URINARY BLADDER FROZEN
Catalog # MSPP-CRL-1473
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
CELL LINE HT-1197 URINARY BLADDER FROZEN
Catalog # MSPP-CRL-1473
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: CRL-1473
Frequently Bought Together
Accessories for eBlot® L1 Fast Wet Transfer System for Mini Gels
Supplier: Genscript
Blot Boxes, IBI Scientific
Supplier: IBI Scientific
10X Tris-Glycine
Supplier: Rockland Immunochemical
OSM (227AA) HUMAN >95% 50UG
Supplier: Genscript
CELL LINE HUMAN URINARY BLADDER 5637
Supplier: 0000013421
Mix & Go! Dilution Buffer, Zymo Research
Supplier: Zymo Research
10X Tris-Acetate-EDTA (TAE)
Supplier: Rockland Immunochemical
Pierce™ Ni-NTA Magnetic Agarose, Thermo Scientific
Supplier: Invitrogen
ScanLater™ Western Blot Assay Kit, Molecular Devices
Supplier: Molecular Devices
AzureSpectra Fluorescent Western Blotting Kits, Azure Biosystems
Supplier: Azure Biosystems
Borate Buffered Saline (BBS), Rockland Immunochemicals
Supplier: Rockland Immunochemical
picoLUCENT™ PLUS ECL Reagent for Western blotting, G-Biosciences
Supplier: G-Biosciences
EGTA 1M PH 8.0 STERILE 100ML
Supplier: 0000014548
BLOK™ Casein, 1% Casein Protein Blocking Solution, G-Biosciences
Supplier: G-Biosciences
FGF-BASIC (154AA) HUMAN >95% 50UG
Supplier: Genscript
Accessories for eZwest™ Lite Automated Western Device
Supplier: Genscript
Borate Saline Buffered (BSB), Rockland Immunochemicals
Supplier: Rockland Immunochemical
100-Place Microscope Slide Box
Supplier: Alkali Scientific