3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
Glass bottle, 25 g
Catalog # TS22928-0250
Supplier: Thermo Scientific Chemicals
Supplier #: 22928-0250
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
Catalog # TS22928-0250
Supplier: Thermo Scientific Chemicals
CAS Number: 54827-17-7
Jump to:
Product Details & Documents
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
- CAS Number:54827-17-7
- Formula:C₁₆H₂₀N₂
- MDL Number:MFCD00007748
- Molecular Weight:240.35 g/mol
- Melting Pt:167…170 °C
- Storage Temperature:Ambient
- EINECS:259-364-6
Specifications
Specification Test Results
- Appearance:White to cream to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:167 to 172 °C
- Loss on drying:≤0.5 %
- Sulfated ash:≤0.2 %
- HPLC:≥99.0 %
- Titration with HClO4:98.0 to 102.0 %
Specifications
- Pack type:Glass bottle
- Size:25 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



