PAN SAMPLE ALUMINUM 6.7MM PK200
Catalog # MSPP-900786901
Supplier: TA INSTRUMENTS INC MS
PAN SAMPLE ALUMINUM 6.7MM PK200
Catalog # MSPP-900786901
Supplier: TA INSTRUMENTS INC MS
Supplier Number: 900786.901
Frequently Bought Together
VWR® Sterile 70% Isopropanol
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
SCIENCEWARE® Large Weighted Safety Shield, Thermo Scientific
Supplier: Bel-Art
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
VWR® Weighing Paper
Supplier: VWR International
Isoprene 99% stabilized
Supplier: Thermo Scientific Chemicals
Paraformaldehyde 97%
Supplier: Thermo Scientific Chemicals