CARBIDE BUR OPR FRICTION GRIP 1/4 PK10
Catalog # MSPP-1007205
Supplier: HENRY SCHEIN MS
CARBIDE BUR OPR FRICTION GRIP 1/4 PK10
Catalog # MSPP-1007205
Supplier: HENRY SCHEIN MS
Supplier Number: 1007205
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
Micro-Touch® NitraFree™ Pink Examination Gloves, Ansell
Supplier: Ansell Healthcare
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Weighing Paper
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
Cover Glasses, Hardvard Apparatus
Supplier: Avantor