POWDER BALANCED FED BATCH CHO FEED 4 10L
Catalog # MSPP-94134-10L
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
POWDER BALANCED FED BATCH CHO FEED 4 10L
Catalog # MSPP-94134-10L
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
Supplier Number: 94134-10L
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
Corning® Polycarbonate Erlenmeyer Flasks with Screw Caps
Supplier: Corning
PROSAT® Presaturated Knitted Polynit Wipes, Contec®
Supplier: Contec
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
Nalgene® Cryogenic Vials, Polypropylene, Sterile, External Thread with Screw Cap, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
High Volume MSL 8 Magnetic Stirrer, Velp Scientific
Supplier: VELP SCIENTIFIC INC.
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
MEDIA BALANCD TRANSFECTORY CHO LIQUD 1L
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
NanoPhotometer® N60-TOUCH UV/Visible Spectrophotometer for NanoVolume Applications, Implen
Supplier: IMPLEN U.S.A. INC