2-(4-Chlorophenylthio)ethanol
Bottle, 1 g
Catalog # 76567-768
Supplier: APOLLO SCIENTIFIC
Supplier #: OR15484-1G
Jump to:
Product Details & Documents
Synonyms: 2-[(4-Chlorophenyl)sulphanyl]-1-ethanol, 4-Chlorophenyl-2-hydroxyethyl sulfide, 2-[(4-Chlorophenyl)sulfanyl]-1-ethanol, 4-Chlorophenyl-2-hydroxyethyl sulphide
- CAS Number:13457-98-2
- Formula:C₈H₉ClOS
- MDL Number:MFCD00014037
- Molecular Weight:188.68
- :Density: 1.07 g/cm³ (20 °C)
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Size:1 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



