2-(4-Chlorophenylthio)ethanol
Bottle, 5 g
76567-770
: OR15484-5G
: 2-[(4-Chlorophenyl)sulphanyl]-1-ethanol, 4-Chlorophenyl-2-hydroxyethyl sulfide, 2-[(4-Chlorophenyl)sulfanyl]-1-ethanol, 4-Chlorophenyl-2-hydroxyethyl sulphide
- :13457-98-2
- :C₈H₉ClOS
- :MFCD00014037
- :188.68
- :Density: 1.07 g/cm³ (20 °C)
- :Ambient
- Pack type:Bottle
- Size:5 g