Sellstrom 812 Series Safety Goggles
Product Details & Documents
Sellstrom’s 812 series safety goggles are made with a soft, flexible PVC body to ensure proper fit and comfort.
The non-vented goggles offer the best protection from liquids and dust particles, while the indirect vent goggles include four white indirect vents that allow maximum airflow and superior ventilation; Indirect vent goggles help protect against liquids, dust, smoke and chemical splashes, allowing you to work in a safe and secure environment.
Made from uv-absorbing polycarbonate, the clear lens provides optical clarity for a wide variety of applications and include an adjustable 18" flame-retardant headband for secure fitting. Choose from a variety of options: Indirect vented/non-vented, uncoated/anti-fog, and 200/160/144 pieces per case. Fits over most prescription eyewear.
Certifications: 812 series safety goggles meets ANSI Z87.1+ high-impact standards. Models S81210 and S81220 are CSA certified.
Specifications
- Lens coating:Anti-Fog
- Lens tint:Clear
- Frame material:PVC
- Lens material:Polycarbonate
- Type:Non-vent