Order Entry
United States
Orders LinkContactUsLinkComponent
Eisco® Push Pull Spring Scales

Eisco® Push Pull Spring Scales

Push/pull spring scale, 250 g/2.5 N, graduation interval: 5 g
Catalog # 470346-114
Supplier:   Wards
Supplier #:  PH0035A
Eisco® Push Pull Spring Scales
Catalog # 470346-114
Supplier:  Wards
Eisco® Push Pull Spring Scales
Eisco® Push Pull Spring Scales

Push/pull spring scale, 250 g/2.5 N, graduation interval: 5 g

Price
$14.75
Log in for availability
Each
Product Details & Documents

Acrylic push/pull balances with different capacities. Can be used to measure both pushing and pulling forces. Ideal for use in classroom for physics experiments and measuring weights of small objects.

  • Transparent acrylic tubular body
  • Spring mechanism is clearly visible
  • Calibrated dual scale

The blue push pull balance (250 g/2.5 N) is designed for measuring force, others allow you to use grams for a mass activity and Newtons for a force activity.

Specifications
  • Description:
    Push/pull spring scale
  • Weighing capacity:
    250 g/2.5 N, graduation interval: 5 g