Order Entry
United States
Orders LinkContactUsLinkComponent
Eisco® Push Pull Spring Scales

Eisco® Push Pull Spring Scales

Push/pull spring scale, 5 kg/50 N, graduation interval: 100 g
Catalog # 470345-574
Supplier:   Wards
Supplier #:  PH0035G
Eisco® Push Pull Spring Scales
Catalog # 470345-574
Supplier:  Wards
Eisco® Push Pull Spring Scales
Eisco® Push Pull Spring Scales

Push/pull spring scale, 5 kg/50 N, graduation interval: 100 g

Price
$23.60
Log in for availability
Each
Product Details & Documents

Acrylic push/pull balances with different capacities. Can be used to measure both pushing and pulling forces. Ideal for use in classroom for physics experiments and measuring weights of small objects.

  • Transparent acrylic tubular body
  • Spring mechanism is clearly visible
  • Calibrated dual scale

The blue push pull balance (250 g/2.5 N) is designed for measuring force, others allow you to use grams for a mass activity and Newtons for a force activity.

Specifications
  • Description:
    Push/pull spring scale
  • Weighing capacity:
    5 kg/50 N, graduation interval: 100 g
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.