Orders LinkContactUsLinkComponent
Eisco Triangular Retort Bases
4", 1.75 lbs.
  470343-870
 :   Wards
 :  CH0648A
Eisco Triangular Retort Bases
  470343-870
 :  Wards
Eisco Triangular Retort Bases
Eisco Triangular Retort Bases
4", 1.75 lbs.
$28.65
Each
Cast iron Triangular Retort Stand with center-tapped hole for maximum stability in minimum footprint.
  • Three-point retort bases made of chemical-resistant cast iron with a black coating
  • Offers maximum stability in a minimal footprint
  • Sturdy construction provides superb support for experimental setups
  • Features a centered-tapped hole for a 10×1.5 mm threaded rod (not included)
Triangular 3-Point Retort Stands are made of chemically-resistant cast iron with a black finish. Features a centered tapped hole suitable for rods with a 10×1.5 mm metric thread. Provides superb stability for basic experiments and setups. The durable and sturdy construction of these bases make them an excellent choice for both science classrooms and professional laboratories.
  • Length:
    4"
  • Weight:
    1.75 lbs.
Accessories
  •  
    Description
    Ø
    Rod length
  • 470345-310
    Steel support rod
    12 mm
    508 mm (20")
    Each - $10.70
  •  
    Description
    Ø
    Rod length
  • 470343-698
    Aluminum support rod
    12 mm
    609 mm (24")
    Each - $12.10
  •  
    Description
    Ø
    Rod length
  • 470345-166
    Steel support rod
    12 mm
    762 mm (30")
    Each - $11.65
  •  
    Description
    Ø
    Rod length
  • 470344-578
    Stainless steel support rod
    12 mm
    762 mm (30")
    Each - $18.25
  •  
    Description
    Ø
    Rod length
  • 470345-558
    Aluminum support rod
    12 mm
    508 mm (20")
    Each - $10.05
  •  
    Description
    Ø
    Rod length
  • 470345-052
    Aluminum support rod
    12 mm
    762 mm (30")
    Each - $16.10
  •  
    Description
    Ø
    Rod length
  • 470344-218
    Stainless steel support rod
    12 mm
    609 mm (24")
    Each - $15.25
  •  
    Description
    Ø
    Rod length
  • 470345-364
    Stainless steel support rod
    12 mm
    508 mm (20")
    Each - $14.30
  •  
    Description
    Ø
    Rod length
  • 470345-394
    Steel support rod
    12 mm
    609 mm (24")
    Each - $14.30
  •  
    Description
    Ø
    Rod length
  • 470345-774
    Stainless steel support rod
    12 mm
    1016 mm (40")
    Each - $24.15
  •  
    Description
    Ø
    Rod length
  • 470345-328
    Aluminum support rod
    12 mm
    1016 mm (40")
    Each - $19.85
  •  
    Description
    Ø
    Rod length
  • 470344-962
    Stainless steel support rod
    12 mm
    901 mm (35.5")
    Each - $24.15