DETECTION KIT PATHHUNTER 100PLT 550ML
Catalog # MSPP-93-0001L
Supplier: Eurofins DiscoverX Corporation
DETECTION KIT PATHHUNTER 100PLT 550ML
Catalog # MSPP-93-0001L
Supplier: Eurofins DiscoverX Corporation
Supplier Number: 93-0001L
Frequently Bought Together
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Vi-CELL BLU Reagent Kits, Beckman Coulter
Supplier: Beckman Coulter
Water, OmniPur®, WFI (water for injection) quality cell culture tested, sterile, Millipore®
Supplier: MilliporeSigma
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
Accessories for Multidrop® Combi Reagent Dispenser, Thermo Scientific
Supplier: Thermo Fisher Scientific
FLIPR® Calcium 5 Fluorescent Assay Kit
Supplier: MOLECULAR DEVICES, LLC MS
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
FLIPR Calcium 6 Assay Kits, Molecular Devices
Supplier: Molecular Devices
Corning® Synthemax® II-SC Substrate, Corning
Supplier: Corning
Screen Quest™ Calbryte-520 Probenecid-Free and Wash-Free Calcium Assay Kit, AAT Bioquest
Supplier: AAT Bioquest
FLIPR® Potassium Assay Kits
Supplier: MOLECULAR DEVICES, LLC MS