Order Entry
United States
Orders LinkContactUsLinkComponent
MicroGenesis Mop

MicroGenesis Mop

MicroGenesis mop frame, blue, 10×44 cm (4×17.5"), PP
Catalog # 76551-222
Supplier:   Contec
Supplier #:  MGMP0009
MicroGenesis Mop
Catalog # 76551-222
Supplier:  Contec
MicroGenesis Mop
MicroGenesis Mop

MicroGenesis mop frame, blue, 10×44 cm (4×17.5"), PP

Price
$193.50
New Customer Promotional Price
Log in for availability
Case of 6 Each
Product Details & Documents

Single-use microfiber mop pads.

  • Synthetic construction is compatible with common disinfectants such as quats, peroxides and bleach
  • New pad for each use simplifies quality control, inventory and space management, all without cross-contamination risk
  • Compatible with most existing hardware

MicroGenesis Mop 100% synthetic, disposable microfiber mop pad effectively removes fine particles and large debris from floors, providing superior removal of visible contaminants. Designed to be used wet or dry for general facilities cleaning in ISO Class 8 environments and above. It is compatible with a wide range of disinfectants, including quats, peroxides and bleach. These synthetic pads are designed to provide uniform surface coverage.

Specifications
  • Description:
    MicroGenesis mop frame, blue
  • Dimensions:
    10×44 cm (4×17.5")
  • Material:
    PP