Order Entry
United States
Orders LinkContactUsLinkComponent
Dropper Assemblies

Dropper Assemblies

Assembly droppers, 2 oz., Glass, 7×89 mm
Catalog # 470338-740
Supplier:   GSC International
Supplier #:  405-4D-GR
Dropper Assemblies
Catalog # 470338-740
Supplier:  GSC International
Dropper Assemblies
Dropper Assemblies

Assembly droppers, 2 oz., Glass, 7×89 mm

Price
$118.20
Log in for availability
Case of 144 Each
Product Details & Documents

We sell many different sizes of dropper assemblies, varying by size, capacity, and the size of threaded neck they are designed to fit.

  • 24/400 neck size
  • Glass pipette with rubber bulb and plastic cap

Straight-talking and practical, our 24/400 neck size glass pipette with rubber bulb and plastic cap is the reliable dropper assembly you need in your lab. This no-frills design features a 20/400 cap that gets the job done without fuss.

The clear, flint glass construction is durable, while the simple rubber bulb and plastic cap provide a secure, leak-proof seal. Whether you're measuring out precious samples or refilling reagents, this dropper assembly puts functionality first. No flashy features, just a solid, trustworthy tool to keep your lab operations running smoothly. Get back to work with this honest, hardworking dropper - no extra bells and whistles required.

Specifications
  • Ø×L:
    7×89 mm
  • Capacity:
    2 oz.
  • Description:
    Assembly droppers
  • Material:
    Glass