Order Entry
United States
Orders LinkContactUsLinkComponent
 

Pipettes

Pipette, 5 ml, 10"
Catalog # 470338-732
Supplier:   GSC International
 :  1601-7-CS
Pipettes
Catalog # 470338-732
Supplier:  GSC International
Pipettes
Pipettes

Pipette, 5 ml, 10"

Price
$302.70
Log in for availability
Case of 120 Each
Product Details & Documents

Used for dispensing liqiuds accurately and quickly.

  • Glass with rubber bulbs

This handy glass tool with a rubber bulb allows you to accurately measure and dispense liquids up to 5 ml. Whether you're doing experiments in the lab or science projects at home, it gives you the control you require. No more messy spills or inaccurate measurements. This simple, yet essential piece of equipment is a must-have for anyone serious about STEM learning and scientific exploration. Stop wasting time and resources with subpar droppers - upgrade to the Pipette for reliable, controlled liquid handling. Your experiments deserve the best, so why settle for anything less?

Specifications
  • Capacity:
    5 ml
  • Description:
    Pipette
  • Length:
    10"