SET I NUCLEOBOND XTRA EF BUFFER
Catalog # MSPP-740427
Supplier: MACHEREY-NAGEL INC MS
SET I NUCLEOBOND XTRA EF BUFFER
Catalog # MSPP-740427
Supplier: MACHEREY-NAGEL INC MS
Supplier Number: 740427
Frequently Bought Together
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Multiwell Cell Culture Plates
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Acetonitrile ≥99.5% for HPLC
Supplier: VWR
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
MIDORI Green Advance Nucleic Acid Staining Solution, Bulldog-bio
Supplier: Bulldog Bio
NUCLEOBOND FINALIZER PLUS
Supplier: Clontech Laboratories