FORWARD PRIMER VIAL MVM KIT 1X 450UL
Catalog # MSPP-M223-1
Supplier: Cygnus
Frequently Bought Together
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Dilux™ Dilution Reservoir
Supplier: DILUX LLC BE
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Quality Economy Brand Luer Lock Syringes, Air-Tite
Supplier: Air-Tite
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
NUCLEASE FREE WATER 2ML FOR MVM KIT
Supplier: Cygnus