Orders LinkContactUsLinkComponent
Forceps, Micro Iris Tissue, Mortech
Forceps, Micro Iris Tissue, Mortech
3.5", Curved
Catalog # 76548-854
Supplier:   Mopec
Supplier #:  AB151
Forceps, Micro Iris Tissue, Mortech
Catalog # 76548-854
Supplier:  Mopec
Forceps, Micro Iris Tissue, Mortech
Forceps, Micro Iris Tissue, Mortech
3.5", Curved
Price
$675.71
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Unique octagon handle shape provides a broad area for the finger to grasp for secure, yet delicate post-mortem dissections.
  • Premium grade stainless steel
  • 1×2 teeth tips
Available with straight or curved edges.
Specifications
  • Length:
    3.5"
  • Curvature:
    Curved