Orders LinkContactUsLinkComponent
Forceps, CutMATE, Mortech
Cutting Forceps, Single, 7", 2 mm blocks
  76548-752
 :   Mopec
 :  AB072
Forceps, CutMATE, Mortech
  76548-752
 :  Mopec
Forceps, CutMATE, Mortech
Forceps, CutMATE, Mortech
Cutting Forceps, Single, 7", 2 mm blocks
$599.88
Each
CutMATE forceps were developed to overcome the variability of tissue specimen thickness. They will allow cutting consistent tissue specimen thickness of 2, 3, or 4 mm.
  • 304 stainless steel
  • Fits all plastic universal cassettes
The unique 'trident-like' design of the 35 mm long ergonomically angled prongs, ensures uniform tissue thickness cutting of tissue lengths that will fit standard cassettes. The inside surface of the prongs is specially textured to provide the extra grip needed for stabilizing and slicing difficult and hard to hold specimens.
  • Description:
    Cutting Forceps, Single
  • Length:
    7"
  • Cutting capacity:
    2 mm blocks