Scissors, Micro Iris, Mortech
Product Details & Documents
Delicate scissors excellent for small, very precise, post-mortem dissection.
- Premium grade stainless steel
- Sharp points
Available with straight or curved edges.
Specifications
- Length:4"
- Curvature:Straight