Order Entry
United States
Orders LinkContactUsLinkComponent
Tissue Marking Dyes, StatLab

Tissue Marking Dyes, StatLab

Margin Marker, Orange, 2 oz
Catalog # 76548-246
Supplier:   StatLab
Supplier #:  SL662OR-2
Tissue Marking Dyes, StatLab
Catalog # 76548-246
Supplier:  StatLab
CAS Number:  
Tissue Marking Dyes, StatLab
Tissue Marking Dyes, StatLab

Margin Marker, Orange, 2 oz

CAS Number:  
Price
$41.22
Log in for availability
Each (2oz)
Product Details & Documents

Permanent tissue marking dyes. Seven colors formulated to define margins without bleeding or fading. Durable, bright dyes hold permanently through processing and staining procedures. Unique lids may either be completely removed or conveniently flipped open to prevent drying.

Set includes one tissue marking dye rack and a set of 2 oz dyes in each of 7 colors.

Packaging: Manufactured in the USA

Specifications

Specifications

  • Color:
    Orange
  • Description:
    Margin Marker
  • Size:
    2 oz

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.