Tissue Marking Dyes, StatLab
Margin Marker, Green, 2 oz
Catalog # 76548-244
Supplier: StatLab
Supplier #: SL662GR-2
Jump to:
Product Details & Documents
Permanent tissue marking dyes. Seven colors formulated to define margins without bleeding or fading. Durable, bright dyes hold permanently through processing and staining procedures. Unique lids may either be completely removed or conveniently flipped open to prevent drying.
Set includes one tissue marking dye rack and a set of 2 oz dyes in each of 7 colors.
Packaging: Manufactured in the USA
Specifications
Specifications
- Color:Green
- Description:Margin Marker
- Size:2 oz



