Eosin, Select
1 gallon
Catalog # 76548-238
Supplier #: SL406
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
STAIN EOSIN SELECT 1GAL Catalog #76548-239
Product 76548-239 is a replacement for 76548-238 but varies by Brand, Packaging or Quantity.
Product Details & Documents
Proprietary, alcoholic and acidified formulation.
- Produces a brilliant cytoplasmic stain with three distinct color variations in a short amount of staining time
- Yields a very familiar cytoplasmic staining result
Part of the StatLab Select stain line.
Manufactured in USA.
Synonyms: Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-
- CAS Number:15086-94-9
- Formula:C₂₀H₈Br₄O₅
- MDL Number:MFCD00036189
- Molecular Weight:647.89 g/mol
- UN:1170
- ADR:3,III
Specifications
Specifications
- Size:1 gallon
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



