Orders LinkContactUsLinkComponent
Eosin, Select
1 pint
  76548-232
 :   StatLab
 :  SL402
Eosin, Select
  76548-232
 :  StatLab
 :  15086-94-9
Eosin, Select
Eosin, Select
1 pint
 :  15086-94-9
$56.90
Each (1pt US)
Proprietary, alcoholic and acidified formulation.
  • Produces a brilliant cytoplasmic stain with three distinct color variations in a short amount of staining time
  • Yields a very familiar cytoplasmic staining result
Part of the StatLab Select stain line.
Manufactured in USA.
 :  Acid Red 87, Tetrabromofluorescein, 2-(3,6-Dihydroxy-2,4,5,7-tetrabromoxanthen-9-yl)-benzoic acid, 2,4,5,7-Tetrabromofluorescein, Eosin yellowish, Solvent red 43, Bromofluorescein, Bromo acid J. TS, XL, or XX, CI 45380, Bronze Bromo ES, Spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one, 2',4',5',7'-tetrabromo-3',6'-dihydroxy-
  •  :15086-94-9
  •  :C₂₀H₈Br₄O₅
  •  :MFCD00036189
  •  :647.89 g/mol
  •  :1170
  •  :3,III
  • Size:
    1 pint