Order Entry
United States
Orders LinkContactUsLinkComponent
Michels Solution , Statlab

Michels Solution , Statlab

Michels Solution, 20 ml
Catalog # 76548-150
Supplier:   StatLab
Supplier #:  MS0507
Michels Solution , Statlab
Catalog # 76548-150
Supplier:  StatLab
CAS Number:  
Michels Solution , Statlab
Michels Solution , Statlab

Michels Solution, 20 ml

CAS Number:  
Price
$231.81
Log in for availability
Case of 96 (20ml)
Product Details & Documents

Michels Solution is used to preserve and transport specimens for examination in immunofluorescence studies.

  • While not a fixative, specimens can be held at room temperature for five days in transport medium before processing
  • This simple salt solution maintains pH, but does not kill most pathogens
  • Specimens received in transport medium should be washed in three changes of Michels Wash Solution (10 minutes for each wash)
  • Not suitable for FISH studies

Part of the StatLab line of formalin alternatives.

Manufactured in the USA

Specifications

Specifications

  • Description:
    Michels Solution
  • Size:
    20 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.