D-(-)-Glutamic acid 99+%
Glass bottle for solids, 25 g
Catalog # TS22789-0250
Supplier: Thermo Scientific Chemicals
Supplier #: 22789-0250
D-(-)-Glutamic acid 99+%
Catalog # TS22789-0250
Supplier: Thermo Scientific Chemicals
CAS Number: 6893-26-1
Jump to:
Product Details & Documents
Synonyms: (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
- CAS Number:6893-26-1
- Formula:C₅H₉NO₄
- MDL Number:MFCD00063112
- Molecular Weight:147.13 g/mol
- Merck Index:13,04482
- Boiling Pt:325 °C (1013 hPa)
- Melting Pt:200…202 °C
- EINECS:230-000-8
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.2 % (105°C, 3 hrs)
- Sulfated ash:≤0.1 %
- HPLC:≤1 % L-content
- Specific optical rotation:-30.5° to -32.2° (20°C, 589 nm) (c=10, 2 N HCl)
- Titration with NaOH:99.0 to 101.0 %
- Other amino acids:≤0.5 % (TLC)
Specifications
- Pack type:Glass bottle for solids
- Size:25 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



